missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dibromopropane, 98%, Thermo Scientific Chemicals

Product Code. 11487676
Click to view available options
Quantity:
250 g
1000 g
5000 g
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

1,3-Dibromopropane, 98%, C3H6Br2, CAS Number-109-64-8, 1,3-dibromo propane, 1,3-dibromo-propane, .alpha.,.gamma.-dibromopropane, alpha,gamma-dibromopropane, ccris 6711, .omega.,.omega.'-dibromopropane, propane, 1,3-dibromo, trimethylene bromide, trimethylene dibromide, unii-yqr3048ix9, 5000g, 635662

1,3-Dibromopropane is used to prepare C3-bridged compounds through C-N coupling reactions. It is used in the preparation of 2,3,4,5-tetrahydro-7,7-diphenylimidazo- [2,1-b]-thiazine-6(7H)-one by reacting with potassium salt of 5,5-diphenyl-2-thiohydantoin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Dibromopropane is used to prepare C3-bridged compounds through C-N coupling reactions. It is used in the preparation of 2,3,4,5-tetrahydro-7,7-diphenylimidazo- [2,1-b]-thiazine-6(7H)-one by reacting with potassium salt of 5,5-diphenyl-2-thiohydantoin.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-64-8
Molecular Formula C3H6Br2
Molecular Weight (g/mol) 201.89
MDL Number MFCD00000255
InChI Key VEFLKXRACNJHOV-UHFFFAOYSA-N
Synonym trimethylene bromide, trimethylene dibromide, propane, 1,3-dibromo, alpha,gamma-dibromopropane, unii-yqr3048ix9, ccris 6711, .alpha.,.gamma.-dibromopropane, 1,3-dibromo propane, 1,3-dibromo-propane, .omega.,.omega.'-dibromopropane
PubChem CID 8001
IUPAC Name 1,3-dibromopropane
SMILES BrCCCBr

Specifications

Melting Point -34°C
Density 1.99
Boiling Point 167°C
Flash Point 54°C (130°F)
Assay Percent Range 98%
Linear Formula Br(CH2)3Br
Refractive Index 1.523
Quantity 1000 g
UN Number UN1993
Beilstein 635662
Merck Index 14,9713
Solubility Information Miscible with water.
Formula Weight 201.9
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1,3-Dibromopropane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'