Learn More
1,3-Dibromopropane, 98%, Thermo Scientific Chemicals
1,3-Dibromopropane, 98%, C3H6Br2, CAS Number-109-64-8, 1,3-dibromo propane, 1,3-dibromo-propane, .alpha.,.gamma.-dibromopropane, alpha,gamma-dibromopropane, ccris 6711, .omega.,.omega.'-dibromopropane, propane, 1,3-dibromo, trimethylene bromide, trimethylene dibromide, unii-yqr3048ix9, 5000g, 635662
£40.10 - £402.00
Chemical Identifiers
CAS | 109-64-8 |
---|---|
Molecular Formula | C3H6Br2 |
Molecular Weight (g/mol) | 201.89 |
MDL Number | MFCD00000255 |
InChI Key | VEFLKXRACNJHOV-UHFFFAOYSA-N |
Synonym | trimethylene bromide, trimethylene dibromide, propane, 1,3-dibromo, alpha,gamma-dibromopropane, unii-yqr3048ix9, ccris 6711, .alpha.,.gamma.-dibromopropane, 1,3-dibromo propane, 1,3-dibromo-propane, .omega.,.omega.'-dibromopropane |
PubChem CID | 8001 |
IUPAC Name | 1,3-dibromopropane |
SMILES | BrCCCBr |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11497676
|
Thermo Scientific Alfa Aesar
A14687.30 |
250 g |
£40.10
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11487676
|
Thermo Scientific Alfa Aesar
A14687.0B |
1000 g |
£105.00
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11407686
|
Thermo Scientific Alfa Aesar
A14687.0I |
5000 g |
£402.00
5000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1,3-Dibromopropane is used to prepare C3-bridged compounds through C-N coupling reactions. It is used in the preparation of 2,3,4,5-tetrahydro-7,7-diphenylimidazo- [2,1-b]-thiazine-6(7H)-one by reacting with potassium salt of 5,5-diphenyl-2-thiohydantoin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,3-Dibromopropane is used to prepare C3-bridged compounds through C-N coupling reactions. It is used in the preparation of 2,3,4,5-tetrahydro-7,7-diphenylimidazo- [2,1-b]-thiazine-6(7H)-one by reacting with potassium salt of 5,5-diphenyl-2-thiohydantoin.
Solubility
Miscible with water.
Notes
Incompatible with strong oxidizing agents and strong bases.
Chemical Identifiers
109-64-8 | |
201.89 | |
VEFLKXRACNJHOV-UHFFFAOYSA-N | |
8001 | |
BrCCCBr |
C3H6Br2 | |
MFCD00000255 | |
trimethylene bromide, trimethylene dibromide, propane, 1,3-dibromo, alpha,gamma-dibromopropane, unii-yqr3048ix9, ccris 6711, .alpha.,.gamma.-dibromopropane, 1,3-dibromo propane, 1,3-dibromo-propane, .omega.,.omega.'-dibromopropane | |
1,3-dibromopropane |
Specifications
109-64-8 | |
1.99 | |
54°C (130°F) | |
C3H6Br2 | |
1.523 | |
UN1993 | |
14,9713 | |
Miscible with water. | |
BrCCCBr | |
201.89 | |
201.9 | |
Liquid |
°C to 34°C | |
167°C | |
98% | |
Br(CH2)3Br | |
MFCD00000255 | |
635662 | |
trimethylene bromide, trimethylene dibromide, propane, 1,3-dibromo, alpha,gamma-dibromopropane, unii-yqr3048ix9, ccris 6711, .alpha.,.gamma.-dibromopropane, 1,3-dibromo propane, 1,3-dibromo-propane, .omega.,.omega.'-dibromopropane | |
VEFLKXRACNJHOV-UHFFFAOYSA-N | |
1,3-dibromopropane | |
8001 | |
98% | |
1, 3-Dibromopropane |
Safety and Handling
GHS H Statement
H226-H302-H315-H319
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
H226-H302-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'einecsNumber' : 203-690-3
missing translation for 'rtecsNumber' : TX8575000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only