missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichloro-2-iodo-5-(trifluoromethyl)benzene, 97%, Thermo Scientific™

Product Code. 10747495
Click to view available options
Quantity:
1 g
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 175205-56-8
Molecular Formula C7H2Cl2F3I
Molecular Weight (g/mol) 340.89
MDL Number MFCD00084997
InChI Key RLCCFAVBZGICHD-UHFFFAOYSA-N
Synonym 1,3-dichloro-2-iodo-5-trifluoromethyl benzene, 3,5-dichloro-4-iodobenzotrifluoride, 3,5-dichloro-4-iodobenztrifluoride, buttpark 96\50-28, benzene,1,3-dichloro-2-iodo-5-trifluoromethyl, 1,3-bis chloranyl-2-iodanyl-5-trifluoromethyl benzene, 3,5-dichloro-4-iodo-alpha,alpha,alpha-trifluorotoluene
PubChem CID 2736838
IUPAC Name 1,3-dichloro-2-iodo-5-(trifluoromethyl)benzene
SMILES FC(F)(F)C1=CC(Cl)=C(I)C(Cl)=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 50 g
Formula Weight 340.89
Percent Purity 97%
Chemical Name or Material 1, 3-Dichloro-2-iodo-5-(trifluoromethyl)benzene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'