missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichlorobenzene, 98%, Thermo Scientific Chemicals

Product Code. 10377800
Click to view available options
Quantity:
5 mL
25 mL
250 mL
500 mL
1 L
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 541-73-1
Molecular Formula C6H4Cl2
Molecular Weight (g/mol) 147
MDL Number MFCD00000573
InChI Key ZPQOPVIELGIULI-UHFFFAOYSA-N
Synonym m-dichlorobenzene, m-phenylene dichloride, m-dichlorobenzol, meta-dichlorobenzene, benzene, 1,3-dichloro, metadichlorobenzene, benzene, m-dichloro, m-phenylenedichloride, m-dcb, 3-dichlorobenzene
PubChem CID 10943
ChEBI CHEBI:36693
IUPAC Name 1,3-dichlorobenzene
SMILES C1=CC(=CC(=C1)Cl)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -24°C
Color Colorless to Yellow
Density 1.2800g/mL
Boiling Point 172°C to 173°C
Flash Point 67°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H4Cl2
Refractive Index 1.5450 to 1.547
Quantity 500 mL
Beilstein 05, 202
Merck Index 15, 3064
Specific Gravity 1.28
Solubility Information Solubility in water: 0.0123g/100mL water (25°C). Other solubilities: soluble in organic solvents such as alcohol,,benzene,ether
Viscosity 1.045 mPa.s (23°C)
Formula Weight 147
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1, 3-Dichlorobenzene, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'