Learn More
1,3-Dichlorotetramethyldisiloxane, 96%, Thermo Scientific Chemicals
£34.47 - £117.59
Chemical Identifiers
CAS | 2401-73-2 |
---|---|
Molecular Formula | C4H12Cl2OSi2 |
Molecular Weight (g/mol) | 203.209 |
MDL Number | MFCD00018088 |
InChI Key | DMEXFOUCEOWRGD-UHFFFAOYSA-N |
Synonym | 1,3-dichloro-1,1,3,3-tetramethyldisiloxane, 1,3-dichlorotetramethyldisiloxane, disiloxane, 1,3-dichloro-1,1,3,3-tetramethyl, chloro chlorodimethylsilyl oxy dimethylsilane, 1,1,3,3-tetramethyl-1,3-dichlorodisiloxane, acmc-209g7w, oxybis chlorodimethylsilane, sym-dichlorotetramethyldisiloxane, dmexfouceowrgd-uhfffaoysa, 1,3-dichlorotetramethyl disiloxane |
PubChem CID | 75461 |
IUPAC Name | chloro-[chloro(dimethyl)silyl]oxy-dimethylsilane |
SMILES | C[Si](C)(O[Si](C)(C)Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11372306
|
Thermo Scientific Alfa Aesar
L05964.06 |
5 g |
£34.47
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11382306
|
Thermo Scientific Alfa Aesar
L05964.14 |
25 g |
£117.59
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1,3-Dichlorotetramethyldisiloxane is used to produce 1,1,3,3-tetramethyl-disiloxane-1,3-diol. It is also used as Chemical Additives. Pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,3-Dichlorotetramethyldisiloxane is used to produce 1,1,3,3-tetramethyl-disiloxane-1,3-diol. It is also used as Chemical Additives. Pharmaceutical intermediates.
Solubility
Reacts with water.
Notes
Moisture Sensitive. Store away from strong bases and water. Protect from water and humidity. Incompatible with acids, bases and oxidizing agents. Avoid excessive heat and light.
Chemical Identifiers
2401-73-2 | |
203.209 | |
DMEXFOUCEOWRGD-UHFFFAOYSA-N | |
75461 | |
C[Si](C)(O[Si](C)(C)Cl)Cl |
C4H12Cl2OSi2 | |
MFCD00018088 | |
1,3-dichloro-1,1,3,3-tetramethyldisiloxane, 1,3-dichlorotetramethyldisiloxane, disiloxane, 1,3-dichloro-1,1,3,3-tetramethyl, chloro chlorodimethylsilyl oxy dimethylsilane, 1,1,3,3-tetramethyl-1,3-dichlorodisiloxane, acmc-209g7w, oxybis chlorodimethylsilane, sym-dichlorotetramethyldisiloxane, dmexfouceowrgd-uhfffaoysa, 1,3-dichlorotetramethyl disiloxane | |
chloro-[chloro(dimethyl)silyl]oxy-dimethylsilane |
Specifications
136°C to 138°C | |
2401-73-2 | |
MFCD00018088 | |
1741218 | |
1,3-dichloro-1,1,3,3-tetramethyldisiloxane, 1,3-dichlorotetramethyldisiloxane, disiloxane, 1,3-dichloro-1,1,3,3-tetramethyl, chloro chlorodimethylsilyl oxy dimethylsilane, 1,1,3,3-tetramethyl-1,3-dichlorodisiloxane, acmc-209g7w, oxybis chlorodimethylsilane, sym-dichlorotetramethyldisiloxane, dmexfouceowrgd-uhfffaoysa, 1,3-dichlorotetramethyl disiloxane | |
C[Si](C)(O[Si](C)(C)Cl)Cl | |
203.209 | |
203.22 | |
1,3-Dichlorotetramethyldisiloxane |
-37°C | |
C4H12Cl2OSi2 | |
UN2985 | |
15°C (59°F) | |
DMEXFOUCEOWRGD-UHFFFAOYSA-N | |
chloro-[chloro(dimethyl)silyl]oxy-dimethylsilane | |
75461 | |
96% |
Safety and Handling
GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: CHLOROSILANES, FLAMMABLE, CORROSIVE, N.O.S.
missing translation for 'tsca' : TSCA
RUO – Research Use Only