missing translation for 'onlineSavingsMsg'
Learn More

1,3-Difluoro-2-nitrobenzene, 98%, Thermo Scientific Chemicals

Product Code. 15432018
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1,3-Difluoro-2-nitrobenzene, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Difluoro-2-nitrobenzene, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19064-24-5
Molecular Formula C6H3F2NO2
Molecular Weight (g/mol) 159.092
MDL Number MFCD00192035
InChI Key SSNCMIDZGFCTST-UHFFFAOYSA-N
Synonym 2,6-difluoronitrobenzene, 1,3-difluoro-2-nitro-benzene, benzene, 1,3-difluoro-2-nitro, 2,6-difluoro nitrobenzene, pubchem4403, 1,3-difluoronitrobenzene, acmc-1buv1, 2,6-difluoro-nitrobenzene, 2,6-difluoro nitro benzene, 1,3-difluoro-2-nitrobenzen
PubChem CID 87922
IUPAC Name 1,3-difluoro-2-nitrobenzene
SMILES C1=CC(=C(C(=C1)F)[N+](=O)[O-])F

Specifications

Melting Point 0°C
Density 1.503
Boiling Point 91°C to 92°C (11 mmHg)
Flash Point 88°C (190°F)
Refractive Index 1.494
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 159.09
Percent Purity 98%
Chemical Name or Material 1,3-Difluoro-2-nitrobenzene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'