missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Dihydroxyacetone dimer, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
1000 g
Description
1,3-Dihydroxyacetone dimer is used as a catalytic agent and petrochemical additive. It is also used as a building block in organic synthesis. Further, it serves as a reagent in three-carbon nucleophilic or electrophilic component in various reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,3-Dihydroxyacetone dimer is used as a catalytic agent and petrochemical additive. It is also used as a building block in organic synthesis. Further, it serves as a reagent in three-carbon nucleophilic or electrophilic component in various reactions.
Solubility
Soluble in ethanol, ethyl ether, acetone. Slightly soluble in water. Insoluble in ligroin.
Notes
Hygroscopic. Store in a cool place. Incompatible with strong oxidizing agents.
1,3-Dihydroxyacetone dimer is used as a catalytic agent and petrochemical additive. It is also used as a building block in organic synthesis. Further, it serves as a reagent in three-carbon nucleophilic or electrophilic component in various reactions.
Solubility
Soluble in ethanol, ethyl ether, acetone. Slightly soluble in water. Insoluble in ligroin.
Notes
Hygroscopic. Store in a cool place. Incompatible with strong oxidizing agents.
Specifications
Specifications
Melting Point | ∼75°C to 82°C |
Quantity | 1000 g |
Beilstein | 112910 |
Merck Index | 14,3182 |
Solubility Information | Soluble in ethanol,ethyl ether,acetone. Slightly soluble in water. Insoluble in ligroin. |
IUPAC Name | 2,5-bis(hydroxymethyl)-1,4-dioxane-2,5-diol |
Formula Weight | 180.16 |
Percent Purity | 97% |
Chemical Name or Material | 1,3-Dihydroxyacetone dimer |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'