missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dihydroxyacetone dimer, 97%, Thermo Scientific Chemicals

Product Code. 11472257
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1,3-Dihydroxyacetone dimer is used as a catalytic agent and petrochemical additive. It is also used as a building block in organic synthesis. Further, it serves as a reagent in three-carbon nucleophilic or electrophilic component in various reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Dihydroxyacetone dimer is used as a catalytic agent and petrochemical additive. It is also used as a building block in organic synthesis. Further, it serves as a reagent in three-carbon nucleophilic or electrophilic component in various reactions.

Solubility
Soluble in ethanol, ethyl ether, acetone. Slightly soluble in water. Insoluble in ligroin.

Notes
Hygroscopic. Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62147-49-3
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.08
MDL Number MFCD00051019
InChI Key RXKJFZQQPQGTFL-UHFFFAOYSA-N
Synonym 1,3-dihydroxyacetone dimer, 1,3-dihydroxypropan-2-one dimer, 2,5-bis hydroxymethyl-1,4-dioxane-2,5-diol, 2,5-dihydroxy-1,4-dioxane-2,5-dimethanol, dihydroxyacetone dimer, 2,5-dihydroxydioxane-2,5-dimethanol, 1,4-dioxane-2,5-dimethanol, 2,5-dihydroxy, acmc-20aoav, pubchem4073
PubChem CID 4180364
SMILES OCC(=O)CO

Specifications

Melting Point ∼75°C to 82°C
Quantity 250 g
Beilstein 112910
Merck Index 14,3182
Solubility Information Soluble in ethanol,ethyl ether,acetone. Slightly soluble in water. Insoluble in ligroin.
IUPAC Name 1,3-dihydroxypropan-2-one
Formula Weight 180.16
Percent Purity 97%
Chemical Name or Material 1,3-Dihydroxyacetone dimer

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'