missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dihydroxynaphthalene, 99+%

Product Code. 10498710 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 10498710

Brand: Thermo Scientific Acros 163340050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 132-86-5 | C10H8O2 | 160.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132-86-5
Molecular Formula C10H8O2
Molecular Weight (g/mol) 160.17
InChI Key XOOMNEFVDUTJPP-UHFFFAOYSA-N
Synonym 1,3-naphthalenediol, 1,3-dihydroxynaphthalene, naphthoresorcinol, naphthoresoucinol, 3-hydroxybenzocyclohexadien-1-one, naphthoresorcin, ccris 7896, unii-5x457yew8y, acmc-1bvwb, naphthalenediol-1,3
PubChem CID 8601
IUPAC Name naphthalene-1,3-diol
SMILES C1=CC=C2C(=C1)C=C(C=C2O)O

Specifications

Melting Point 123.0°C to 126.0°C
Color Brown to Orange-Pink
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula C10H6(OH)2
Quantity 5 g
Beilstein 06,978
Merck Index 15,6481
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and ether,soluble in ethanol an diethyl ether,slightly soluble in acetone,benzene and ligroin
Formula Weight 160.17
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 1, 3-Dihydroxynaphthalene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.