missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethyl-1H-pyrazole-5-carbaldehyde, ≥90%, Thermo Scientific™

Product Code. 10485502
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 25016-09-5
Molecular Formula C6H8N2O
Molecular Weight (g/mol) 124.143
MDL Number MFCD02681907
InChI Key JWYFGNVBKRJGTN-UHFFFAOYSA-N
PubChem CID 2776362
IUPAC Name 2,5-dimethylpyrazole-3-carbaldehyde
SMILES CC1=NN(C(=C1)C=O)C

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 1,3-dimethyl-1h-pyrazole-5-carbaldehyde, 1,3-dimethylpyrazole-5-carbaldehyde, 1h-pyrazole-5-carboxaldehyde, 1,3-dimethyl, 1,3-dimethyl-5-formyl-1h-pyrazole, 2,5-dimethyl-2h-pyrazole-3-carbaldehyde, 1,3-dimethylpyrazole-5-carboxaldehyde, 1,3-dimethyl-1h-pyrazole-5-carboxaldehyde, 2,5-dimethyl-3-pyrazolecarboxaldehyde, 1,3-dimethyl-1h-pyrazol-5-carbaldehyde, 1,3-dimethyl-1 h-pyrazole-5-carbaldehyde
Formula Weight 124.14
Percent Purity 90%
Chemical Name or Material 1, 3-Dimethyl-1H-pyrazole-5-carbaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'