missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethyl-1H-thieno[2,3-c]pyrazole-5-carbonyl chloride, 97%, Thermo Scientific™

Product Code. 10522862
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 388088-75-3
Molecular Formula C8H7ClN2OS
Molecular Weight (g/mol) 214.67
MDL Number MFCD03086153
InChI Key BQOTYHLFKDBAGL-UHFFFAOYSA-N
Synonym 1,3-dimethyl-1h-thieno 2,3-c pyrazole-5-carbonyl chloride, 1,3-dimethylthieno 2,3-c pyrazole-5-carbonyl chloride, 1,3-dimethyl-1h-thieno 2,3-c pyrazole-5-carbonylchloride, 1,3-dimethylthiopheno 3,2-d pyrazole-5-carbonyl chloride, 1h-thieno 2,3-c pyrazole-5-carbonylchloride, 1,3-dimethyl
PubChem CID 2776542
IUPAC Name 1,3-dimethylthieno[2,3-c]pyrazole-5-carbonyl chloride
SMILES CN1N=C(C)C2=C1SC(=C2)C(Cl)=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 214.68
Percent Purity 97%
Chemical Name or Material 1, 3-Dimethyl-1H-thieno[2, 3-c]pyrazole-5-carbonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'