missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethyl-5-morpholino-1H-pyrazol-4-amine, 97%, Thermo Scientific™

Product Code. 10177352
Amber Glass Bottle; 1g
Click to view available options
Amber Glass Bottle; 1g
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 568577-87-7
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C9H16N4O
MDL Number MFCD04115380
Quantity 1 g
Synonym 1,3-dimethyl-5-morpholino-1h-pyrazol-4-amine, 1,3-dimethyl-5-morpholin-4-yl pyrazol-4-amine, 4-amino-1,3-dimethyl-5-morpholino-4-yl-pyrazole, 1,3-dimethyl-5-morpholin-4-ylpyrazole-4-ylamine, 1,3-dimethyl-5-morpholin-4-yl-1h-pyrazol-4-amine
InChI Key LEVZXNJICHTQEC-UHFFFAOYSA-N
SMILES CC1=NN(C(=C1N)N2CCOCC2)C
IUPAC Name 1,3-dimethyl-5-morpholin-4-ylpyrazol-4-amine
Molecular Weight (g/mol) 196.254
PubChem CID 2795380
Formula Weight 196.25
Percent Purity 97%
Chemical Name or Material 1, 3-Dimethyl-5-morpholino-1H-pyrazol-4-amine
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,3-Dimethyl-5-morpholino-1H-pyrazol-4-amine, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.