missing translation for 'onlineSavingsMsg'
Learn More
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide hydrochloride, 98+%, Thermo Scientific Chemicals Product Code.: 11419742

1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide hydrochloride, 98+%, Thermo Scientific Chemicals

Product Code. 11419742
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11419742

Brand: Thermo Scientific Alfa Aesar A10807.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide hydrochloride is used as a carboxyl activating agent. It is used in peptide synthesis, 3'-amino-3'-deoxyadenosine-5'-di- and triphosphates and in the preparation of antibodies like immunoconjugates. It plays a vital role for immobilization of large biomolecules in association with N-hydroxysuccinimide. It is also used in the acylation of phosphoranes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide hydrochloride is used as a carboxyl activating agent. It is used in peptide synthesis, 3′-amino-3′-deoxyadenosine-5′-di- and triphosphates and in the preparation of antibodies like immunoconjugates. It plays a vital role for immobilization of large biomolecules in association with N-hydroxysuccinimide. It is also used in the acylation of phosphoranes.

Solubility
Soluble in acetonitrile dichloromethane, dimethylformamide, and tetrahydrofuran. Slightly soluble in water.

Notes
Sensitive to moisture. Incompatible with strong acids, strong oxidizing agents and moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25952-53-8
Molecular Formula C8H18ClN3
Molecular Weight (g/mol) 191.70
MDL Number MFCD00012503
InChI Key FPQQSJJWHUJYPU-UHFFFAOYSA-N
Synonym 1-3-dimethylaminopropyl-3-ethylcarbodiimide hydrochloride, edc.hcl, edci, edc hydrochloride, wsc hcl, edac hydrochloride, edac, hydrochloride, 1-ethyl-3-3-dimethylaminopropyl carbodiimide hydrochloride, edcl, edac hclShow MoreShow Less
PubChem CID 2723939
IUPAC Name 3-(ethyliminomethylideneamino)-N,N-dimethylpropan-1-amine;hydrochloride
SMILES Cl.CCN=C=NCCCN(C)C

Specifications

Melting Point ∽113°C
Linear Formula CH3CH2N=C=N(CH2)3N(CH3)2·HCl
Quantity 5 g
Beilstein 5764110
Sensitivity Moisture sensitive
Solubility Information Soluble in acetonitrile dichloromethane,dimethylformamide,and tetrahydrofuran. Slightly soluble in water.
Formula Weight 191.7
Percent Purity ≥98%
Chemical Name or Material 1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide hydrochloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10253350 Certificate TypeCertificate of Analysis Date11/12/2024 Catalog NumberA10807.03,A10807.14,A10807.06
Lot NumberS12K035 Certificate TypeCertificate of Analysis Date19/06/2024 Catalog NumberA10807.03,A10807.14,A10807.06
Lot Number10233296 Certificate TypeCertificate of Analysis Date25/01/2024 Catalog NumberA10807.03,A10807.14,A10807.06
Lot Number10233295 Certificate TypeCertificate of Analysis Date11/01/2024 Catalog NumberA10807.03,A10807.14,A10807.06
Lot NumberN24J035 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog NumberA10807.03,A10807.14,A10807.06
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1-(3-Dimethylaminopropyl)-3-ethylcarbodiimidehydrochloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : N'-(ethylcarbonimidoyl)-N,N-dimethyl-1,3-Propanediamine monohydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide hydrochloride, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.