missing translation for 'onlineSavingsMsg'
Learn More
1,3-Dimethylbarbituric acid, 99% (dry wt.), water <6%, Thermo Scientific Chemicals Product Code.: 11424674

1,3-Dimethylbarbituric acid, 99% (dry wt.), water <6%, Thermo Scientific Chemicals

Product Code. 11424674
250 g
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy

Product Code. 11424674

Brand: Thermo Scientific Alfa Aesar A13692.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

1,3-Dimethylbarbituric acid is used as a catalyst in the Knoevenagel condensation of a series of aromatic aldehydes. It is also used in the synthesis of 5-aryl-6-(alkyl- or aryl-amino)-1,3-dimethylfuro [2,3-d]pyrimidine derivatives and enantioselective synthesis of isochromene pyrimidinedione derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 769-42-6
Molecular Formula C6H8N2O3
Molecular Weight (g/mol) 156.141
MDL Number MFCD00006675
InChI Key VVSASNKOFCZVES-UHFFFAOYSA-N
Synonym 1,3-dimethylbarbituric acid, n,n'-dimethylbarbituric acid, 2,4,6 1h,3h,5h-pyrimidinetrione, 1,3-dimethyl, 1,3-dimethylbarbituricacid, barbituric acid, 1,3-dimethyl, 1,3-dimethylpyrimidine-2,4,6 1h,3h,5h-trione, 1,3-dimethyl barbituric acid, 1,3-dimethyl-2,4,6 1h,3h,5h-pyrimidinetrione, 1,3-dimethyl-2,4,6 1h,3h,5h pyrimidinetrione, 1,3-dimethyl-1,3,5-trihydropyrimidine-2,4,6-trioneShow MoreShow Less
PubChem CID 69860
IUPAC Name 1,3-dimethyl-1,3-diazinane-2,4,6-trione
SMILES CN1C(=O)CC(=O)N(C1=O)C

Specifications

Melting Point 121°C to 123°C
Beilstein 139810
Formula Weight 156.14
Percent Purity 99%
Concentration or Composition (by Analyte or Components) water <6%
Quantity 250 g
Physical Form Crystalline Powder
Assay (dry weight)
Chemical Name or Material 1,3-Dimethylbarbituric acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10251053 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA13692.0B,A13692.30,A13692.18
Lot Number10239674 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA13692.0B,A13692.30,A13692.18
Lot Number10239675 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA13692.0B,A13692.30,A13692.18
Lot Number10233161 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13692.0B,A13692.30,A13692.18
Lot Number10227652 Certificate TypeCertificate of Analysis Date07/08/2020 Catalog NumberA13692.0B,A13692.30,A13692.18
compliance-icons compliance-icons
Product Identifier
  • 1,3-Dimethylbarbituric acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 1,3-Dimethylbarbituric acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,3-Dimethylbarbituric acid, 99% (dry wt.), water <6%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.