missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1,3-Dioxolan-2-ylmethyl)zinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
50 mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Reacts with water.
Notes
Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Reacts with water.
Notes
Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Spécification
Spécification
CAS | 1135389-59-1 |
Density | 0.992 |
Molecular Formula | C4H7BrO2Zn |
MDL Number | MFCD07698709 |
Quantity | 50 mL |
UN Number | UN3399 |
Sensitivity | Air sensitive |
Synonym | bromo 1,3-dioxolan-2-ylmethyl zinc, 1,3-dioxolan-2-ylmethyl zinc bromide, lojrlqrgbkthdb-uhfffaoysa-m, 1,3-dioxolan-2-ylmethyl zinc bromide, 0.6 m in thf, 1,3-dioxolan-2-ylmethyl zinc bromide, 0.50 m in thf, 1,3-dioxolan-2-ylmethyl zinc bromide 0.5 m in tetrahydrofuran, 1,3-dioxolan-2-ylmethyl zinc bromide, 0.5m in thf, packaged under argon in resealable chemseal bottles |
Solubility Information | Reacts with water. |
InChI Key | ZEBRGOCCZOTNDU-UHFFFAOYSA-M |
Afficher plus |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'