missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dioxolane, 99.5+%, pure, stabilized, Thermo Scientific Chemicals

Product Code. 10628134
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
Glass Bottle
Metal drum

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 646-06-0, 128-37-0
Molecular Formula C3H6O2
Molecular Weight (g/mol) 74.08
MDL Number MFCD00003207
InChI Key WNXJIVFYUVYPPR-UHFFFAOYSA-N
Synonym dioxolane, glycolformal, formal glycol, 1,3-dioxolan, 1,3-dioxacyclopentane, 1,3-dioxalane, ethylene glycol formal, glycol methylene ether, glycol formal, dioxolan czech
PubChem CID 31404
ChEBI CHEBI:34247
IUPAC Name 2,6-ditert-butyl-4-methylphenol
SMILES CC1=CC(=C(C(=C1)C(C)(C)C)O)C(C)(C)C

Specifications

CAS Min % 99.4
CAS Max % 100.0
Melting Point -95°C
Color White
Density 1.0600g/mL
Boiling Point 74.0°C to 75.0°C
Flash Point −6°C
Infrared Spectrum Authentic
Assay Percent Range 99.5% min. (GC)
Packaging Metal drum
Refractive Index 1.3990 to 1.4010
Quantity 25 L
Beilstein 19, 2
Fieser 14,281
Specific Gravity 1.06
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,ether and acetone
Stabilizer 75ppm BHT max.
Viscosity 0.66 mPa.s (20°C)
Formula Weight 74.08
Percent Purity 99.5+%
Grade Pure
Physical Form Crystalline Powder
Chemical Name or Material 1, 3-Dioxolane, Stabilized
Show More Show Less
compliance-icons
Product Identifier
  • 1,3-Dioxolane, stabilized
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'