missing translation for 'onlineSavingsMsg'
Learn More
1,3-Diphenyl-1,3-propanedione, 98+%, Thermo Scientific Chemicals Product Code.: 11455513

1,3-Diphenyl-1,3-propanedione, 98+%, Thermo Scientific Chemicals

Product Code. 11455513
100 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11455513

Brand: Thermo Scientific Alfa Aesar A13450.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1,3-Diphenyl-1,3-propanedione, is a minor constituent of licorice found to inhibit mammary tumorigenesis, lymphomas, and leukemias in mice. It acts as a heat stabilizer& antitumour agent. It is used in perfumery and as well as in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Diphenyl-1,3-propanedione, is a minor constituent of licorice found to inhibit mammary tumorigenesis, lymphomas, and leukemias in mice. It acts as a heat stabilizer& antitumour agent. It is used in perfumery and as well as in organic synthesis.

Solubility
It is Soluble in ether, chloroform, and aqueous sodium hydroxide. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents, Acid anhydrides, Halogens, halogen acids. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-46-7
Molecular Formula C15H12O2
Molecular Weight (g/mol) 224.259
MDL Number MFCD00003085
InChI Key NZZIMKJIVMHWJC-UHFFFAOYSA-N
Synonym dibenzoylmethane, 1,3-diphenyl-1,3-propanedione, 2-benzoylacetophenone, phenyl phenacyl ketone, 1,3-propanedione, 1,3-diphenyl, rhodiastab 83, omega-benzoylacetophenone, dibenzoyl-methane, karenzu dk2, unii-ans7me8okcShow MoreShow Less
PubChem CID 8433
ChEBI CHEBI:75417
IUPAC Name 1,3-diphenylpropane-1,3-dione
SMILES C1=CC=C(C=C1)C(=O)CC(=O)C2=CC=CC=C2

Specifications

Melting Point 76°C to 80°C
Boiling Point 219°C to 221°C (18 mmHg)
Quantity 100 g
Beilstein 514910
Merck Index 14,3009
Solubility Information It is Soluble in ether,chloroform,and aqueous sodium hydroxide. Insoluble in water.
Formula Weight 224.26
Percent Purity ≥98%
Chemical Name or Material 1,3-Diphenyl-1,3-propanedione

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10244690 Certificate TypeCertificate of Analysis Date08/12/2022 Catalog NumberA13450.14,A13450.36,A13450.22
Lot Number10236515 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13450.14,A13450.36,A13450.22
Lot Number10155864 Certificate TypeCertificate of Analysis Date07/08/2020 Catalog NumberA13450.14,A13450.36,A13450.22
Lot Number10211743 Certificate TypeCertificate of Analysis Date07/08/2020 Catalog NumberA13450.14,A13450.36,A13450.22
Lot Number10220367 Certificate TypeCertificate of Analysis Date07/08/2020 Catalog NumberA13450.14,A13450.36,A13450.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,3-Diphenyl-1,3-propanedione, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.