missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Ethoxycarbonylphenyl)pyrrole, 98%, Thermo Scientific™

Product Code. 15446317
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83140-93-6
Molecular Formula C13H13NO2
Molecular Weight (g/mol) 215.252
MDL Number MFCD06205096
InChI Key BOAJVZUYGSTURO-UHFFFAOYSA-N
Synonym ethyl 3-pyrrol-1-yl benzoate, ethyl 3-1-pyrrolyl benzoate, ethyl 3-1-pyrrolyl-benzoate, 1-3-ethoxycarbonylphenyl pyrrole, 3-1-pyrrolyl benzoic acid ethyl ester
PubChem CID 4763724
IUPAC Name ethyl 3-pyrrol-1-ylbenzoate
SMILES CCOC(=O)C1=CC(=CC=C1)N2C=CC=C2

Specifications

Melting Point 64°C to 65°C
Boiling Point 122°C (0.19 mmHg)
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 215.25
Percent Purity 98%
Chemical Name or Material 1-(3-Ethoxycarbonylphenyl)pyrrole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'