missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Propanediol, 98%, Thermo Scientific Chemicals
£54.27 - £1054.25
Chemical Identifiers
CAS | 504-63-2 |
---|---|
Molecular Formula | C3H8O2 |
Molecular Weight (g/mol) | 76.10 |
MDL Number | MFCD00002949 |
InChI Key | YPFDHNVEDLHUCE-UHFFFAOYSA-N |
Synonym | 1,3-propanediol, trimethylene glycol, 1,3-dihydroxypropane, 2-deoxyglycerol, 1,3-propylene glycol, 1,3-propylenediol, 1,3-propandiol, 2-hydroxymethyl ethanol, omega-propanediol, beta-propylene glycol |
PubChem CID | 10442 |
ChEBI | CHEBI:16109 |
IUPAC Name | propane-1,3-diol |
SMILES | OCCCO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10765781
|
Thermo Scientific Acros
131402500 |
250 mL | Glass bottle |
£54.27
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10041410
|
Thermo Scientific Acros
131400010 |
1 L | Glass bottle |
£150.75
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11964631
|
Thermo Scientific Acros
131400025 |
2.5 L | Glass bottle |
£334.67
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15159173
|
Thermo Scientific Acros
131400100 |
10 L | Metal drum |
£1054.25
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identificatori chimici
504-63-2 | |
76.10 | |
YPFDHNVEDLHUCE-UHFFFAOYSA-N | |
10442 | |
propane-1,3-diol |
C3H8O2 | |
MFCD00002949 | |
1,3-propanediol, trimethylene glycol, 1,3-dihydroxypropane, 2-deoxyglycerol, 1,3-propylene glycol, 1,3-propylenediol, 1,3-propandiol, 2-hydroxymethyl ethanol, omega-propanediol, beta-propylene glycol | |
CHEBI:16109 | |
OCCCO |
Specifica
504-63-2 | |
1.0520g/mL | |
140°C | |
97.5% min. (GC) | |
HO(CH2)3OH | |
MFCD00002949 | |
13,268 | |
1.052 | |
Solubility in water: 100g/L water. Other solubilities: soluble in ethanol, diethyl ether,, slightly soluble in benzene | |
OCCCO | |
76.10 | |
CHEBI:16109 | |
76.09 | |
Oily Liquid |
-32.0°C | |
214.0°C | |
Authentic | |
C3H8O2 | |
1.4380 to 1.4400 | |
01,475 | |
15,9885 | |
1,3-propanediol, trimethylene glycol, 1,3-dihydroxypropane, 2-deoxyglycerol, 1,3-propylene glycol, 1,3-propylenediol, 1,3-propandiol, 2-hydroxymethyl ethanol, omega-propanediol, beta-propylene glycol | |
YPFDHNVEDLHUCE-UHFFFAOYSA-N | |
propane-1,3-diol | |
10442 | |
52.7 mPa.s (20°C) | |
98% | |
1, 3-Propanediol |
Sicurezza e movimentazione
missing translation for 'einecsNumber' : 207-997-3
RUO – Research Use Only