missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-[3-(tert-Butyl)phenyl]hydrazine hydrochloride 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
Specifications
Quantity | 250 mg |
Formula Weight | 200.71 |
Percent Purity | Tech. |
Grade | Technical |
Chemical Name or Material | 1-[3-(tert-Butyl)phenyl]hydrazine hydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction