missing translation for 'onlineSavingsMsg'
Learn More

1,3-Thiazol-2-ylmethanol, 97+%, Thermo Scientific™

Product Code. 10376964
Click to view available options
Quantity:
1 g
5 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 14542-12-2
Molecular Formula C4H5NOS
Molecular Weight (g/mol) 115.15
MDL Number MFCD06200855
InChI Key JNHDLNXNYPLBMJ-UHFFFAOYSA-N
Synonym thiazol-2-ylmethanol, 2-thiazolemethanol, thiazol-2-yl-methanol, 2-hydroxymethylthiazole, 2-hydroxymethyl thiazole, 2-hydroxymehtylthiazole, 1,3-thiazol-2-yl methanol, thiazol-2-yl methanol, thiazole-2-methanol, 2-thiazolylmethanol
PubChem CID 2795213
SMILES OCC1=NC=CS1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
IUPAC Name 1,3-thiazol-2-ylmethanol
Formula Weight 115.16
Percent Purity 97%
Chemical Name or Material 1, 3-Thiazol-2-ylmethanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'