missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Thiazol-5-ylmethylamine hydrochloride, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
IUPAC Name | 1,3-thiazol-5-ylmethanamine;hydrochloride |
Formula Weight | 187.09 |
Percent Purity | 97% |
Chemical Name or Material | 1, 3-Thiazol-5-ylmethylamine di-hydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction