missing translation for 'onlineSavingsMsg'
Learn More

1-[3-(Trifluoromethyl)phenylsulfonylamino]cyclohexanecarboxylic acid, 96%, Thermo Scientific™

Product Code. 15458895
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 690645-95-5
Molecular Formula C14H16F3NO4S
Molecular Weight (g/mol) 351.34
MDL Number MFCD05866546
InChI Key DDEMDTVJTHGHDA-UHFFFAOYSA-N
Synonym 1-3-trifluoromethyl phenyl sulfonamido cyclohexane-1-carboxylic acid, 1-3-trifluoromethyl phenylsulfonylamino cyclohexanecarboxylic acid, 1-3-trifluoromethyl benzenesulfonamido cyclohexane-1-carboxylic acid, 1-3-trifluoromethyl phenyl sulfonylamino cyclohexane-1-carboxylic acid, 1-3-trifluoromethyl phenylsulfonamido cyclohexanecarboxylic acid
PubChem CID 2241577
IUPAC Name 1-[[3-(trifluoromethyl)phenyl]sulfonylamino]cyclohexane-1-carboxylic acid
SMILES C1CCC(CC1)(C(=O)O)NS(=O)(=O)C2=CC=CC(=C2)C(F)(F)F

Specifications

Melting Point 164°C to 165°C
Quantity 250 mg
Formula Weight 351.35
Percent Purity 96%
Chemical Name or Material 1-[3-(Trifluoromethyl)phenylsulfonylamino]cyclohexanecarboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'