Learn More
Trajan Scientific and Medical™ 1/32 in. O.D. PEEKsil™ Tubing
A mechanically robust polymer-sheathed fused silica tubing for use in HPLC systems
Brand: Trajan Scientific and Medical™ 0624319
This item is not returnable.
View return policy
Description
- Fused silica internal wall provides smooth surface, free of imperfections
- PEEKsil compatible with most organic solvents and resistant to strong acids
- Effective pH range of 0 to 10
- Flexibility ensures easy connection between columns, detectors and injectors
Specifications
Tubing | |
2/Pk. | |
0.1 mm | |
500 mm |
Product Content Correction
Product Title
Trajan Scientific and Medical™ 1/32 in. O.D. PEEKsil™ Tubing > 0.1mm I.D.; 500 mm L; 2/pack
Spot an opportunity for improvement?Share a Content Correction