missing translation for 'onlineSavingsMsg'
Learn More

1-[4-(4-Morpholinyl)phenyl]guanidine, 98%, Thermo Scientific Chemicals

Product Code. 15446577
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 247234-41-9
Molecular Formula C11H16N4O
Molecular Weight (g/mol) 220.28
MDL Number MFCD11986905
InChI Key ZVZJREQBRCRGLM-UHFFFAOYSA-N
Synonym 1-4-morpholinophenyl guanidine, n-4-morpholin-4-ylphenyl guanidine, 4-morpholinophenylguanidine, 4-morpholino phenylguanidine, n-4-morpholin-4-yl-phenyl-guanidine, n-4-morpholin-4-yl phenyl guanidine, 1-4-4-morpholinyl phenyl guanidine, 1-4-morpholin-4-yl phenyl guanidine, 2-4-morpholin-4-yl phenyl guanidine
PubChem CID 10176830
IUPAC Name 2-(4-morpholin-4-ylphenyl)guanidine
SMILES NC(N)=NC1=CC=C(C=C1)N1CCOCC1

Specifications

Melting Point ∼220°C (decomposition)
Density 1.309
Quantity 5 g
Formula Weight 220.27
Percent Purity 98%
Chemical Name or Material 1-[4-(4-Morpholinyl)phenyl]guanidine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'