missing translation for 'onlineSavingsMsg'
Learn More

1,4,7,10-Tetraazacyclotridecane, Thermo Scientific Chemicals

Product Code. 15473517
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1,4,7,10-Tetraazacyclotridecane is used to find the binding constants, and for building block elements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4,7,10-Tetraazacyclotridecane is used to find the binding constants, and for building block elements.

Solubility
Soluble in water.

Notes
Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 295-14-7
Molecular Formula C9H22N4
Molecular Weight (g/mol) 186.303
MDL Number MFCD06410986
InChI Key LADZJJOUGVGJHM-UHFFFAOYSA-N
Synonym 1,4,7,10-tetraazacyclotridecane, homocyclen, acmc-20ap3n, 13 anen4
PubChem CID 11478723
IUPAC Name 1,4,7,10-tetrazacyclotridecane
SMILES C1CNCCNCCNCCNC1

Specifications

Quantity 250 mg
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 186.3
Chemical Name or Material 1,4,7,10-Tetraazacyclotridecane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'