missing translation for 'onlineSavingsMsg'
Learn More

1,4,8,11-Tetraazacyclotetradecane, 98%, Thermo Scientific Chemicals

£55.20 - £649.00

Chemical Identifiers

CAS 295-37-4
Molecular Formula C10H24N4
Molecular Weight (g/mol) 200.33
MDL Number MFCD00005105
InChI Key MDAXKAUIABOHTD-UHFFFAOYSA-N
Synonym 1,4,8,11-tetraazacyclotetradecane, cyclam, 14 anen4, 1,4,8,11-tetraazacyclo-tetradecane, 1,4,8,11-tetraazacyclotetradecane cyclam, ips2, acmc-209h9n, 5-26-11-00027 beilstein handbook reference, 1,8,11-tetraazacyclotetradecane, 1,4,8,11tetraaza-cyclotetradecane
PubChem CID 64964
ChEBI CHEBI:37401
IUPAC Name 1,4,8,11-tetrazacyclotetradecane
SMILES C1CNCCNCCCNCCNC1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10125210
View Documents Promotion Details
Thermo Scientific Acros
269330010
1 g Glass bottle
£55.20
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10075863
View Documents Promotion Details
Thermo Scientific Acros
269330050
5 g Glass bottle
£185.00
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
13448498
View Documents Promotion Details
Thermo Scientific Acros
269330250
25 g Glass Bottle
£649.00
25g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent
Specifications

Chemical Identifiers

295-37-4
200.33
MDAXKAUIABOHTD-UHFFFAOYSA-N
64964
1,4,8,11-tetrazacyclotetradecane
C10H24N4
MFCD00005105
1,4,8,11-tetraazacyclotetradecane, cyclam, 14 anen4, 1,4,8,11-tetraazacyclo-tetradecane, 1,4,8,11-tetraazacyclotetradecane cyclam, ips2, acmc-209h9n, 5-26-11-00027 beilstein handbook reference, 1,8,11-tetraazacyclotetradecane, 1,4,8,11tetraaza-cyclotetradecane
CHEBI:37401
C1CNCCNCCCNCCNC1

Specifications

295-37-4
100.0
White
97.5% min. (GC)
MFCD00005105
17,89
Solubility in water: 5g/100mL (20°C).
C1CNCCNCCCNCCNC1
200.33
CHEBI:37401
98%
1,4,8,11-Tetraazacyclotetradecane
99.0
185.0°C to 188.0°C
Authentic
C10H24N4
26,1647
1,4,8,11-tetraazacyclotetradecane, cyclam, 14 anen4, 1,4,8,11-tetraazacyclo-tetradecane, 1,4,8,11-tetraazacyclotetradecane cyclam, ips2, acmc-209h9n, 5-26-11-00027 beilstein handbook reference, 1,8,11-tetraazacyclotetradecane, 1,4,8,11tetraaza-cyclotetradecane
MDAXKAUIABOHTD-UHFFFAOYSA-N
1,4,8,11-tetrazacyclotetradecane
64964
200.33
Fluffy Needle-like Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 206-039-1

Product Identifier
  • 1,4,8,11-Tetraazacyclotetradecane

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only