missing translation for 'onlineSavingsMsg'
Learn More
1,4,8-Tri-Boc-1,4,8,11-tetraazacyclotetradecane Product Code.: 15457615

1,4,8-Tri-Boc-1,4,8,11-tetraazacyclotetradecane

Product Code. 15457615
250 mg
Click to view available options
Quantity:
250 mg
This item is not returnable. View return policy

Product Code. 15457615

Brand: Thermo Scientific Alfa Aesar H26741.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 170161-27-0 | C25H48N4O6 | 500.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 48°C to 54°C
CAS 170161-27-0
Molecular Formula C25H48N4O6
MDL Number MFCD09263315
Synonym 1,4,8-tri-boc-1,4,8,11-tetraazacyclotetradecane, tri-tert-butyl 1,4,8,11-tetraazacyclotetradecane-1,4,8-tricarboxylate, acmc-1c98h, 1,4,8-tri-t-butyloxycarbonyl-1,4,8,11-tetraazacyclotetradecane, 1,4,8,11-tetraazacyclotetradecane-1,4,8-tricarboxylic acid tri-tert-butyl ester, 1,4,8,11-tetraazacyclotetradecane-1,4,8-tricarboxylic acid tris 1,1-dimethylethyl ester, 1,4,8-tri-tert-butyl 1,4,8,11-tetraazacyclotetradecane-1,4,8-tricarboxylate
InChI Key FIPOUUYFPSMVMX-UHFFFAOYSA-N
SMILES CC(C)(C)OC(=O)N1CCCN(CCN(CCCNCC1)C(=O)OC(C)(C)C)C(=O)OC(C)(C)C
IUPAC Name tritert-butyl 1,4,8,11-tetrazacyclotetradecane-1,4,8-tricarboxylate
Molecular Weight (g/mol) 500.68
Quantity 250 mg
PubChem CID 10940041
Formula Weight 500.68
Sensitivity Very Hygroscopic
Chemical Name or Material 1,4,8-Tri-Boc-1,4,8,11-tetraazacyclotetradecane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,4,8-Tri-Boc-1,4,8,11-tetraazacyclotetradecane >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.