missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Acetylpiperidino)ethan-1-one, 97%, Thermo Scientific™

Product Code. 10321473
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 162368-01-6
Molecular Formula C9H15NO2
Molecular Weight (g/mol) 169.22
MDL Number MFCD00106774
InChI Key VFZXZEJGLKOTBW-UHFFFAOYSA-N
Synonym 1-4-acetylpiperidino ethan-1-one, 1-1-acetylpiperidin-4-yl ethanone, 1,4-diacetylpiperidine, 1-4-acetylpiperidin-1-yl ethan-1-one, 1,1'-piperidine-1,4-diyl diethanone, 1-1-acetyl-piperidin-4-yl-ethanone, piperidine, 1,4-diacetyl-9ci, 1-4-acetylpiperidin-1-yl ethanone, ethanone,1-1-acetyl-4-piperidinyl, piperidine, 1,4-diacetyl
PubChem CID 2777250
IUPAC Name 1-(1-acetylpiperidin-4-yl)ethanone
SMILES CC(=O)C1CCN(CC1)C(C)=O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 169.22
Percent Purity 97%
Chemical Name or Material 1-(4-Acetylpiperidino)ethan-1-one
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.