missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Anthraquinone, 94%, Thermo Scientific Chemicals
£63.60 - £248.00
Chemical Identifiers
CAS | 635-12-1 |
---|---|
Molecular Formula | C14H8O2 |
Molecular Weight (g/mol) | 208.216 |
MDL Number | MFCD00013724 |
InChI Key | LSOTZYUVGZKSHR-UHFFFAOYSA-N |
Synonym | 1,4-anthraquinone, 1,4-anthracenedione, 1,4-dioxoanthracene, 1,4-anthracenequinone, acmc-1b3md, 1,4-dihydroanthracene-1,4-dione |
PubChem CID | 69457 |
IUPAC Name | anthracene-1,4-dione |
SMILES | C1=CC=C2C=C3C(=O)C=CC(=O)C3=CC2=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11330114
|
Thermo Scientific Alfa Aesar
L00573.06 |
5 g |
£63.60
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11340114
|
Thermo Scientific Alfa Aesar
L00573.14 |
25 g |
£248.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1,4-Anthraquinone is used in the preparation of 1-methyl-1,4,4a,12a-tetrahydronaphthacene-5,12-dione.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,4-Anthraquinone is used in the preparation of 1-methyl-1,4,4a,12a-tetrahydronaphthacene-5,12-dione.
Solubility
Insoluble in water.
Notes
Incompatible with oxidizing agents.
Chemical Identifiers
635-12-1 | |
208.216 | |
LSOTZYUVGZKSHR-UHFFFAOYSA-N | |
69457 | |
C1=CC=C2C=C3C(=O)C=CC(=O)C3=CC2=C1 |
C14H8O2 | |
MFCD00013724 | |
1,4-anthraquinone, 1,4-anthracenedione, 1,4-dioxoanthracene, 1,4-anthracenequinone, acmc-1b3md, 1,4-dihydroanthracene-1,4-dione | |
anthracene-1,4-dione |
Specifications
635-12-1 | |
C14H8O2 | |
1871197 | |
Insoluble in water. | |
C1=CC=C2C=C3C(=O)C=CC(=O)C3=CC2=C1 | |
208.216 | |
208.22 | |
1,4-Anthraquinone |
∽218°C (decomposition) | |
MFCD00013724 | |
1,4-anthraquinone, 1,4-anthracenedione, 1,4-dioxoanthracene, 1,4-anthracenequinone, acmc-1b3md, 1,4-dihydroanthracene-1,4-dione | |
LSOTZYUVGZKSHR-UHFFFAOYSA-N | |
anthracene-1,4-dione | |
69457 | |
94% |
Safety and Handling
missing translation for 'einecsNumber' : 211-228-7
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only