Learn More
1,4-Benzenediboronic acid, 96%, Thermo Scientific Chemicals
£31.40 - £330.00
Chemical Identifiers
CAS | 4612-26-4 |
---|---|
Molecular Formula | C6H8B2O4 |
Molecular Weight (g/mol) | 165.746 |
MDL Number | MFCD00236018 |
InChI Key | BODYVHJTUHHINQ-UHFFFAOYSA-N |
Synonym | 1,4-benzenediboronic acid, 1,4-phenylenediboronic acid, 1,4-phenylenebisboronic acid, benzene-1,4-diboronic acid, 4-boronophenyl boronic acid, p-benzenediboronic acid, p-phenylenediboronic acid, 1,4-phenylenebis boronic acid, 1,4 phenyl diboronic acid, boronic acid, 1,4-phenylenebis |
PubChem CID | 230478 |
IUPAC Name | (4-boronophenyl)boronic acid |
SMILES | B(C1=CC=C(C=C1)B(O)O)(O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11471047
|
Thermo Scientific Alfa Aesar
B24064.03 |
1 g |
£31.40
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11481047
|
Thermo Scientific Alfa Aesar
B24064.06 |
5 g |
£88.90
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11491047
|
Thermo Scientific Alfa Aesar
B24064.14 |
25 g |
£330.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used for palladium-catalyzed sequential alkenylation and conjugate addition reactions, scholl cyclizations, Suzuki-Miyaura cross-coupling reactions. It is also used in the preparation of indolizine derivatives as OLEDs, in organic thin-film transistors, Fluorescence and solution-processable coordination polymers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used for palladium-catalyzed sequential alkenylation and conjugate addition reactions, scholl cyclizations, Suzuki-Miyaura cross-coupling reactions. It is also used in the preparation of indolizine derivatives as OLEDs, in organic thin-film transistors, Fluorescence and solution-processable coordination polymers.
Solubility
Soluble in water 2.5%.
Notes
Store in a cool, dry place in tightly closed container. Store at room temperature. Incompatible with oxidizing agents.
Chemical Identifiers
4612-26-4 | |
165.746 | |
BODYVHJTUHHINQ-UHFFFAOYSA-N | |
230478 | |
B(C1=CC=C(C=C1)B(O)O)(O)O |
C6H8B2O4 | |
MFCD00236018 | |
1,4-benzenediboronic acid, 1,4-phenylenediboronic acid, 1,4-phenylenebisboronic acid, benzene-1,4-diboronic acid, 4-boronophenyl boronic acid, p-benzenediboronic acid, p-phenylenediboronic acid, 1,4-phenylenebis boronic acid, 1,4 phenyl diboronic acid, boronic acid, 1,4-phenylenebis | |
(4-boronophenyl)boronic acid |
Specifications
4612-26-4 | |
C6H8B2O4 | |
2836921 | |
Soluble in water 2.5%. | |
B(C1=CC=C(C=C1)B(O)O)(O)O | |
165.746 | |
165.75 | |
1,4-Benzenediboronic acid |
>300°C | |
MFCD00236018 | |
1,4-benzenediboronic acid, 1,4-phenylenediboronic acid, 1,4-phenylenebisboronic acid, benzene-1,4-diboronic acid, 4-boronophenyl boronic acid, p-benzenediboronic acid, p-phenylenediboronic acid, 1,4-phenylenebis boronic acid, 1,4 phenyl diboronic acid, boronic acid, 1,4-phenylenebis | |
BODYVHJTUHHINQ-UHFFFAOYSA-N | |
(4-boronophenyl)boronic acid | |
230478 | |
96% |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only