missing translation for 'onlineSavingsMsg'
Learn More

1,4-Benzodioxan-5-carboxylic acid, 98%, Thermo Scientific™

Product Code. 10776085
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. Rückgaberichtlinie anzeigen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Chemische Identifikatoren

CAS 4442-53-9
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.16
MDL Number MFCD00239415
InChI Key VCLSWKVAHAJSFL-UHFFFAOYSA-N
Synonym 1,4-benzodioxan-5-carboxylic acid, 2,3-dihydrobenzo b 1,4 dioxine-5-carboxylic acid, 2,3-dihydro-1,4-benzodioxin-5-carboxylic acid, 1,4-benzodioxane-5-carboxylic acid, 2,3-dihydro-benzo 1,4 dioxine-5-carboxylic acid, 2,3-ethylenedioxybenzoic acid, 2,3-dihydro-1,4-benzodioxane-5-carboxylic acid, 2,3-dihydrobenzo 1,4 dioxine-5-carboxylic acid, 1,4-benzodioxin-5-carboxylic acid, 2,3-dihydro, 2h,3h-benzo e 1,4-dioxin-5-carboxylic acid
PubChem CID 78184
IUPAC Name 2,3-dihydro-1,4-benzodioxine-5-carboxylic acid
SMILES C1COC2=C(C=CC=C2O1)C(=O)O

Spezifikation

Melting Point 90°C to 94°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 180.16
Percent Purity 98%
Chemical Name or Material 1, 4-Benzodioxan-5-carboxylic acid
compliance-icons
Produktidentifikator
  • 1,4-Benzodioxan-5-carboxylic acid
Signalwort
  • Warning
Gefahrenkategorie
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Sicherheitshinweis
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'