missing translation for 'onlineSavingsMsg'
Learn More
1,4-Benzodioxan-6-carboxaldehyde, 98%, Thermo Scientific™ Product Code.: 10633995

1,4-Benzodioxan-6-carboxaldehyde, 98%, Thermo Scientific™

Product Code. 10633995
5 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10633995

Brand: Thermo Scientific Acros 398210050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 29668-44-8
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00010092
InChI Key CWKXDPPQCVWXAG-UHFFFAOYSA-N
Synonym 1,4-benzodioxan-6-carboxaldehyde, 1,4-benzodioxan-6-carbaldehyde, 2,3-dihydrobenzo b 1,4 dioxine-6-carbaldehyde, 1,4-benzodioxane-6-carboxaldehyde, 3,4-ethylenedioxybenzaldehyde, 1,4-benzodioxin-6-carboxaldehyde, 2,3-dihydro, 7-formyl-1,4-benzodioxane, 1,4-benzodioxane-6-aldehyde, 2,3-dihydro-1,4-benzodioxin-6-carbaldehyde, 1,4-benzodioxane-6-carbaldehyde
PubChem CID 248127
IUPAC Name 2,3-dihydro-1,4-benzodioxine-6-carbaldehyde
SMILES C1COC2=C(O1)C=CC(=C2)C=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 52.0°C to 53.0°C
Color Beige-Yellow
Boiling Point 105°C (20.0 hPa)
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 164.16
Percent Purity 98%
Physical Form Chunks and/or Powder
Chemical Name or Material 1, 4-Benzodioxan-6-carboxaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 1,4-Benzodioxan-6-carboxaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,4-Benzodioxan-6-carboxaldehyde, 98%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.