missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Bis(4-pyridylmethyl)benzene, 97%, Thermo Scientific Chemicals
£114.00
Chemical Identifiers
CAS | 52469-20-2 |
---|---|
Molecular Formula | C18H16N2 |
Molecular Weight (g/mol) | 260.34 |
MDL Number | MFCD23379844 |
InChI Key | FFHRKYGEMWVANQ-UHFFFAOYSA-N |
Synonym | 4-4-pyridin-4-ylmethyl phenyl methyl pyridine, 4,4'-p-xylylene bispyridine, 1,4-bis 4-pyridylmethyl benzene, 1,4-bis pyridin-4-ylmethyl benzene, 4,4'-1,4-phenylenebis methylene dipyridine, pyridine, 4,4'-1,4-phenylenebis methylene bis |
PubChem CID | 20646252 |
IUPAC Name | 4-[[4-(pyridin-4-ylmethyl)phenyl]methyl]pyridine |
SMILES | C(C1=CC=NC=C1)C1=CC=C(CC2=CC=NC=C2)C=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15407787
|
Thermo Scientific Alfa Aesar
H37485.06 |
5 g | |||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
52469-20-2 | |
260.34 | |
FFHRKYGEMWVANQ-UHFFFAOYSA-N | |
20646252 | |
C(C1=CC=NC=C1)C1=CC=C(CC2=CC=NC=C2)C=C1 |
C18H16N2 | |
MFCD23379844 | |
4-4-pyridin-4-ylmethyl phenyl methyl pyridine, 4,4'-p-xylylene bispyridine, 1,4-bis 4-pyridylmethyl benzene, 1,4-bis pyridin-4-ylmethyl benzene, 4,4'-1,4-phenylenebis methylene dipyridine, pyridine, 4,4'-1,4-phenylenebis methylene bis | |
4-[[4-(pyridin-4-ylmethyl)phenyl]methyl]pyridine |
Specifications
52469-20-2 | |
C18H16N2 | |
4-4-pyridin-4-ylmethyl phenyl methyl pyridine, 4,4'-p-xylylene bispyridine, 1,4-bis 4-pyridylmethyl benzene, 1,4-bis pyridin-4-ylmethyl benzene, 4,4'-1,4-phenylenebis methylene dipyridine, pyridine, 4,4'-1,4-phenylenebis methylene bis | |
C(C1=CC=NC=C1)C1=CC=C(CC2=CC=NC=C2)C=C1 | |
260.34 | |
260.34 | |
Solid |
White to Yellow | |
MFCD23379844 | |
FFHRKYGEMWVANQ-UHFFFAOYSA-N | |
4-[[4-(pyridin-4-ylmethyl)phenyl]methyl]pyridine | |
20646252 | |
97% | |
1,4-Bis(4-pyridylmethyl)benzene |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only