Learn More
1-(4-Bromobenzyl)-5-methyl-1H-1,2,3-triazole-4-carboxylic acid, 97%, Thermo Scientific™
£48.35 - £289.00
Chemical Identifiers
CAS | 845885-94-1 |
---|---|
Molecular Formula | C11H10BrN3O2 |
Molecular Weight (g/mol) | 296.124 |
MDL Number | MFCD06659088 |
InChI Key | FJFFHBYCFJNIAV-UHFFFAOYSA-N |
Synonym | 1-4-bromobenzyl-5-methyl-1h-1,2,3-triazole-4-carboxylic acid, 1-4-bromophenyl methyl-5-methyl-1,2,3-triazole-4-carboxylic acid, 1-4-bromophenyl methyl-5-methyl-1h-1,2,3-triazole-4-carboxylic acid, 1h-1,2,3-triazole-4-carboxylicacid, 1-4-bromophenyl methyl-5-methyl, 1-4-bromophenyl methyl-5-methyl-4-triazolecarboxylic acid, 1-4-bromophenyl methyl-5-methyltriazole-4-carboxylic acid, 1-4-bromo-benzyl-5-methyl-1h-1,2,3 triazole-4-carboxylic acid |
PubChem CID | 2794828 |
IUPAC Name | 1-[(4-bromophenyl)methyl]-5-methyltriazole-4-carboxylic acid |
SMILES | CC1=C(N=NN1CC2=CC=C(C=C2)Br)C(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10278812
|
Thermo Scientific Maybridge
MO07389CB |
250 mg |
£48.35
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10288812
|
Thermo Scientific Maybridge
MO07389DA |
1 g |
£96.60
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10022464
|
Thermo Scientific Maybridge
MO07389DE |
5 g |
£289.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
845885-94-1 | |
296.124 | |
FJFFHBYCFJNIAV-UHFFFAOYSA-N | |
2794828 | |
CC1=C(N=NN1CC2=CC=C(C=C2)Br)C(=O)O |
C11H10BrN3O2 | |
MFCD06659088 | |
1-4-bromobenzyl-5-methyl-1h-1,2,3-triazole-4-carboxylic acid, 1-4-bromophenyl methyl-5-methyl-1,2,3-triazole-4-carboxylic acid, 1-4-bromophenyl methyl-5-methyl-1h-1,2,3-triazole-4-carboxylic acid, 1h-1,2,3-triazole-4-carboxylicacid, 1-4-bromophenyl methyl-5-methyl, 1-4-bromophenyl methyl-5-methyl-4-triazolecarboxylic acid, 1-4-bromophenyl methyl-5-methyltriazole-4-carboxylic acid, 1-4-bromo-benzyl-5-methyl-1h-1,2,3 triazole-4-carboxylic acid | |
1-[(4-bromophenyl)methyl]-5-methyltriazole-4-carboxylic acid |
Specifications
845885-94-1 | |
97% | |
C11H10BrN3O2 | |
1-4-bromobenzyl-5-methyl-1h-1,2,3-triazole-4-carboxylic acid, 1-4-bromophenyl methyl-5-methyl-1,2,3-triazole-4-carboxylic acid, 1-4-bromophenyl methyl-5-methyl-1h-1,2,3-triazole-4-carboxylic acid, 1h-1,2,3-triazole-4-carboxylicacid, 1-4-bromophenyl methyl-5-methyl, 1-4-bromophenyl methyl-5-methyl-4-triazolecarboxylic acid, 1-4-bromophenyl methyl-5-methyltriazole-4-carboxylic acid, 1-4-bromo-benzyl-5-methyl-1h-1,2,3 triazole-4-carboxylic acid | |
CC1=C(N=NN1CC2=CC=C(C=C2)Br)C(=O)O | |
296.124 | |
296.12 | |
1-(4-Bromobenzyl)-5-methyl-1H-1, 2, 3-triazole-4-carboxylic acid |
97.0 | |
Amber Glass Bottle | |
MFCD06659088 | |
FJFFHBYCFJNIAV-UHFFFAOYSA-N | |
1-[(4-bromophenyl)methyl]-5-methyltriazole-4-carboxylic acid | |
2794828 | |
97% |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable solid.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep container tightly closed.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning