missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(4-Bromophenoxy)hexane, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
50 g
Specifications
Specifications
Assay Percent Range | 97% |
Quantity | 1 g |
Formula Weight | 257.17 |
Chemical Name or Material | 1-(4-Bromophenoxy)hexane |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction