missing translation for 'onlineSavingsMsg'
Learn More
1-(4-Bromophenyl)-3,5-dimethyl-1H-pyrazole, 97%, Thermo Scientific™ Product Code.: 10346324

1-(4-Bromophenyl)-3,5-dimethyl-1H-pyrazole, 97%, Thermo Scientific™

Product Code. 10346324
250 mg, Amber Glass Bottle
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10346324

Brand: Thermo Scientific Maybridge CC68210CB

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 62546-27-4
Molecular Formula C11H11BrN2
Molecular Weight (g/mol) 251.127
MDL Number MFCD09817558
InChI Key ATCMNDCOEJOOSF-UHFFFAOYSA-N
Synonym 1-4-bromophenyl-3,5-dimethyl-1h-pyrazole, 1-4-bromophenyl-3,5-dimethylpyrazole, 1h-pyrazole, 1-4-bromophenyl-3,5-dimethyl, acmc-209n4v
PubChem CID 24229761
IUPAC Name 1-(4-bromophenyl)-3,5-dimethylpyrazole
SMILES CC1=CC(=NN1C2=CC=C(C=C2)Br)C

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 251.13
Percent Purity 97%
Chemical Name or Material 1-(4-Bromophenyl)-3, 5-dimethyl-1H-pyrazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-(4-Bromophenyl)-3,5-dimethyl-1H-pyrazole, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.