missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Butanedio L, 99+%, Thermo Scientific™
Chemical Identifiers
CAS | 110-63-4 |
---|---|
Molecular Formula | C4H10O2 |
Molecular Weight (g/mol) | 90.12 |
MDL Number | MFCD00002968 |
InChI Key | WERYXYBDKMZEQL-UHFFFAOYSA-N |
IUPAC Name | butane-1,4-diol |
SMILES | OCCCCO |
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
110-63-4 | |
90.12 | |
WERYXYBDKMZEQL-UHFFFAOYSA-N | |
OCCCCO |
C4H10O2 | |
MFCD00002968 | |
butane-1,4-diol |
Specifications
110-63-4 | |
229.2°C (1013.0 hPa) | |
99+% | |
C4H10O2 | |
1.4442 to 1.4462 | |
01, 478 | |
1.01 | |
WERYXYBDKMZEQL-UHFFFAOYSA-N | |
butane-1,4-diol | |
70 mPa.s (24°C) | |
1,4-Butanediol |
1.0100 g/mL | |
135°C | |
Glass bottle | |
HO(CH2)4OH | |
MFCD00002968 | |
15, 1518 | |
Solubility aq. soln.: miscible. Other solubilities: soluble in alcohol, slightly soluble in ether | |
OCCCCO | |
90.12 | |
90.12 |
Safety and Handling
GHS H Statement Harmful if swallowed.
May cause drowsiness or dizziness.
GHS P Statement IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 203-786-5
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 1,4-Butanediol
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Specific target organ toxicity Category 3
Hazard Statement
- H336-May cause drowsiness or dizziness.
- H302-Harmful if swallowed.
Precautionary Statement
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only