missing translation for 'onlineSavingsMsg'
Learn More

1,4-Butanediol diacrylate, 85+%, stab. with 50-105 ppm hydroquinone, Thermo Scientific Chemicals

Product Code. 11303018
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1,4-Butanediol diacrylate is used as a multifunctional crosslinker. It is used in drying technology for paintings, coatings, inks, resins and adhesives. It serves as an intermediate product for polymer synthesis in the chemical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1070-70-8
Molecular Formula C10H14O4
Molecular Weight (g/mol) 198.22
MDL Number MFCD00014940
InChI Key JHWGFJBTMHEZME-UHFFFAOYSA-N
Synonym 1,4-butanediol diacrylate, butylene diacrylate, butanediol diacrylate, tetramethylene glycol diacrylate, 1,4-butylene glycol diacrylate, tetramethylene acrylate, tetramethylene diacrylate, 1,4-butylene diacrylate, 1,4-butanediyl diacrylate, 1,4-butanediyl bisacrylate
PubChem CID 70613
SMILES C=CC(=O)OCCCCOC(=O)C=C

Specifications

Color Colorless
Density 1.051 g/mL
Boiling Point 83°C (0.3 mmHg)
Flash Point >110°C (230°F)
Odor Pleasant
Linear Formula CH2=CHCO2(CH2)4O2CCH=CH2
Refractive Index 1.456
Quantity 100 g
UN Number UN1760
Beilstein 1777487
Solubility Information Immiscible with water.
IUPAC Name 4-prop-2-enoyloxybutyl prop-2-enoate
Formula Weight 198.22
Percent Purity ≥85%
Physical Form Liquid
Assay (Reactive Esters)
Chemical Name or Material 1,4-Butanediol diacrylate, Stabilized with 50 to 105 ppm hydroquinone
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.