missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Chlorophenyl)-3-[4-chloro-3-(trifluoromethyl)phenyl]urea, 97%, Thermo Scientific Chemicals

Product Code. 15449616
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 369-77-7
Molecular Formula C14H9Cl2F3N2O
Molecular Weight (g/mol) 349.13
MDL Number MFCD00867294
InChI Key ZFSXZJXLKAJIGS-UHFFFAOYSA-N
Synonym cloflucarban, halocarban, irgasan cf3, cloflucarbon, trifluoromethyldichlorocarbanilide, unii-i5zzy3dc5g, cloflucarban usan, 4,4'-dichloro-3-trifluoromethyl carbanilide, i5zzy3dc5g, halocarban inn
PubChem CID 9719
IUPAC Name 1-(4-chlorophenyl)-3-[4-chloro-3-(trifluoromethyl)phenyl]urea
SMILES FC(F)(F)C1=CC(NC(=O)NC2=CC=C(Cl)C=C2)=CC=C1Cl

Specifications

Quantity 1 g
Formula Weight 349.13
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-(4-Chlorophenyl)-3-[4-chloro-3-(trifluoromethyl)phenyl]urea

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'