missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Cyclohexanedicarboxylic acid, 99+%, mixture of cis and trans, Thermo Scientific Chemicals
£12.90 - £81.90
Chemical Identifiers
CAS | 1076-97-7 |
---|---|
Molecular Formula | C8H12O4 |
Molecular Weight (g/mol) | 172.18 |
MDL Number | MFCD00001465 |
InChI Key | PXGZQGDTEZPERC-UHFFFAOYSA-N |
Synonym | 1,4-cyclohexanedicarboxylic acid, trans-1,4-cyclohexanedicarboxylic acid, cis-1,4-cyclohexanedicarboxylic acid, cis-cyclohexane-1,4-dicarboxylic acid, trans-cyclohexane-1,4-dicarboxylic acid, hexahydroterephthalic acid, trans-hexahydroterephthalic acid, trans-1,4-cyclohexanedicarboxybic acid, cis-hexahydroterephthalic acid, 1,4-dicarboxycyclohexane |
PubChem CID | 14106 |
IUPAC Name | cyclohexane-1,4-dicarboxylic acid |
SMILES | C1CC(CCC1C(=O)O)C(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10556782
|
Thermo Scientific Acros
406040050 |
5 g | Glass bottle |
£12.90
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10741052
|
Thermo Scientific Acros
406041000 |
100 g | Glass bottle |
£29.90
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10310674
|
Thermo Scientific Acros
406045000 |
500 g | Glass bottle | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1076-97-7 | |
172.18 | |
PXGZQGDTEZPERC-UHFFFAOYSA-N | |
14106 | |
C1CC(CCC1C(=O)O)C(=O)O |
C8H12O4 | |
MFCD00001465 | |
1,4-cyclohexanedicarboxylic acid, trans-1,4-cyclohexanedicarboxylic acid, cis-1,4-cyclohexanedicarboxylic acid, cis-cyclohexane-1,4-dicarboxylic acid, trans-cyclohexane-1,4-dicarboxylic acid, hexahydroterephthalic acid, trans-hexahydroterephthalic acid, trans-1,4-cyclohexanedicarboxybic acid, cis-hexahydroterephthalic acid, 1,4-dicarboxycyclohexane | |
cyclohexane-1,4-dicarboxylic acid |
Specifications
1076-97-7 | |
100.0 | |
White | |
99% min. (GC) | |
MFCD00001465 | |
Solubility in water: soluble | |
C1CC(CCC1C(=O)O)C(=O)O | |
172.18 | |
172.18 | |
Powder |
99.0 | |
165°C to 168°C | |
Authentic | |
C8H12O4 | |
1,4-cyclohexanedicarboxylic acid, trans-1,4-cyclohexanedicarboxylic acid, cis-1,4-cyclohexanedicarboxylic acid, cis-cyclohexane-1,4-dicarboxylic acid, trans-cyclohexane-1,4-dicarboxylic acid, hexahydroterephthalic acid, trans-hexahydroterephthalic acid, trans-1,4-cyclohexanedicarboxybic acid, cis-hexahydroterephthalic acid, 1,4-dicarboxycyclohexane | |
PXGZQGDTEZPERC-UHFFFAOYSA-N | |
cyclohexane-1,4-dicarboxylic acid | |
14106 | |
99+% | |
1, 4-Cyclohexanedicarboxylic acid, Mixture of cis and trans |
Safety and Handling
missing translation for 'einecsNumber' : 214-068-6
RUO – Research Use Only