missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Cyclohexanediol, cis + trans, 98%, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
100 g
Beschreibung
1,4-Cyclohexanediol, cis + trans, is used as a chemical, organic, pharmaceutical intermediate. It is also used as an important intermediate for manufacturing of anti-Cancer and anti-Infective Drugs - dihydroartemisinin and dihydroartemisitene dimers. It is also used in co polyester resins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,4-Cyclohexanediol, cis + trans, is used as a chemical, organic, pharmaceutical intermediate. It is also used as an important intermediate for manufacturing of anti-Cancer and anti-Infective Drugs - dihydroartemisinin and dihydroartemisitene dimers. It is also used in co polyester resins.
Solubility
It is highly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, acid chlorides, acid anhydrides, strong acids.
1,4-Cyclohexanediol, cis + trans, is used as a chemical, organic, pharmaceutical intermediate. It is also used as an important intermediate for manufacturing of anti-Cancer and anti-Infective Drugs - dihydroartemisinin and dihydroartemisitene dimers. It is also used in co polyester resins.
Solubility
It is highly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, acid chlorides, acid anhydrides, strong acids.
Spezifikation
Spezifikation
Melting Point | 98°C to 103°C |
Boiling Point | 149°C to 151°C (20 mmHg) |
Flash Point | 65°C (149°F) |
Quantity | 100 g |
Beilstein | 2036658 |
Solubility Information | It is highly soluble in water. |
Formula Weight | 116.16 |
Percent Purity | 98% |
Chemical Name or Material | 1,4-Cyclohexanediol, cis + trans |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'