missing translation for 'onlineSavingsMsg'
Learn More

1,4-Cyclohexanedione monoethylene acetal, 97%, Thermo Scientific Chemicals

£25.43 - £147.74

Chemical Identifiers

CAS 4746-97-8
Molecular Formula C8H12O3
Molecular Weight (g/mol) 156.181
MDL Number MFCD00010214
InChI Key VKRKCBWIVLSRBJ-UHFFFAOYSA-N
Synonym 1,4-dioxaspiro 4.5 decan-8-one, 1,4-cyclohexanedione monoethylene acetal, 1,4-cyclohexanedione monoethylene ketal, 1,4-cyclohexanedione monoethyleneacetal, 1,4-dioxa-spiro 4.5 decan-8-one, 1,4-cyclohexanedione monoethyleneketal, cyclohexane-1,4-dione monoethylene ketal, 1,4-dioxaspiro 4.5 decane-8-one
PubChem CID 567415
IUPAC Name 1,4-dioxaspiro[4.5]decan-8-one
SMILES C1CC2(CCC1=O)OCCO2
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11428603
View Documents
Thermo Scientific Alfa Aesar
A10408.06
5 g
£25.43
5g
Please to purchase this item. Need a web account? Register with us today!
11438603
View Documents
Thermo Scientific Alfa Aesar
A10408.14
25 g
£56.68
25g
Please to purchase this item. Need a web account? Register with us today!
11448603
View Documents
Thermo Scientific Alfa Aesar
A10408.22
100 g
£147.74
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

1,4-Cyclohexanedione is used in the preparation of series of potent analgesic compounds. 1,4-Cyclohexanedione is also used as a building block in the synthesis of tritium labelled probes for the autoradiography study of the dopamine reuptake complex.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Cyclohexanedione is used in the preparation of series of potent analgesic compounds. 1,4-Cyclohexanedione is also used as a building block in the synthesis of tritium labelled probes for the autoradiography study of the dopamine reuptake complex.

Solubility
Soluble in chloroform & methanol.

Notes
Moisture Sensitive. Store away from moisture and oxidizing agents. Incompatible with strong oxidizing agents and strong acids. Ensure adequate ventilation during use.
Specifications

Chemical Identifiers

4746-97-8
156.181
VKRKCBWIVLSRBJ-UHFFFAOYSA-N
567415
C1CC2(CCC1=O)OCCO2
C8H12O3
MFCD00010214
1,4-dioxaspiro 4.5 decan-8-one, 1,4-cyclohexanedione monoethylene acetal, 1,4-cyclohexanedione monoethylene ketal, 1,4-cyclohexanedione monoethyleneacetal, 1,4-dioxa-spiro 4.5 decan-8-one, 1,4-cyclohexanedione monoethyleneketal, cyclohexane-1,4-dione monoethylene ketal, 1,4-dioxaspiro 4.5 decane-8-one
1,4-dioxaspiro[4.5]decan-8-one

Specifications

4746-97-8
C8H12O3
117631
1,4-dioxaspiro 4.5 decan-8-one, 1,4-cyclohexanedione monoethylene acetal, 1,4-cyclohexanedione monoethylene ketal, 1,4-cyclohexanedione monoethyleneacetal, 1,4-dioxa-spiro 4.5 decan-8-one, 1,4-cyclohexanedione monoethyleneketal, cyclohexane-1,4-dione monoethylene ketal, 1,4-dioxaspiro 4.5 decane-8-one
VKRKCBWIVLSRBJ-UHFFFAOYSA-N
1,4-dioxaspiro[4.5]decan-8-one
567415
97%
72°C to 75°C
MFCD00010214
Moisture sensitive
Soluble in chloroform & methanol.
C1CC2(CCC1=O)OCCO2
156.181
156.18
1,4-Cyclohexanedione monoethylene acetal
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 000-000-0

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only