Learn More
1,4-Di-tert-butylbenzene, 98%, Thermo Scientific Chemicals
£33.06 - £125.63
Chemical Identifiers
CAS | 1012-72-2 |
---|---|
Molecular Formula | C14H22 |
Molecular Weight (g/mol) | 190.33 |
MDL Number | MFCD00008836 |
InChI Key | OOWNNCMFKFBNOF-UHFFFAOYSA-N |
Synonym | 1,4-di-tert-butylbenzene, p-di-tert-butylbenzene, benzene, p-di-tert-butyl, benzene, 1,4-bis 1,1-dimethylethyl, unii-55ptx4kh71, bis 1,1-dimethylethyl benzene, benzene, bis 1,1-dimethylethyl, 1,4-di-t-butylbenzene, acmc-2097up, 1,4-bis tert-butyl benzeneShow More |
PubChem CID | 13895 |
IUPAC Name | 1,4-ditert-butylbenzene |
SMILES | CC(C)(C)C1=CC=C(C=C1)C(C)(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11443367
|
Thermo Scientific Alfa Aesar
A17146.06 |
5 g |
£33.06
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11453367
|
Thermo Scientific Alfa Aesar
A17146.14 |
25 g |
£125.63
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1,4-di-tert-butylbenzene (DTBB) is used as a crystallized form from several organic solvents to study the effect of solvents and crystallization conditions on its habit. It is also used as a solvent and an intermediate in the manufacture of other organic compounds for the finished products of such as curing agents, engineering plastics and cross-linking agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,4-di-tert-butylbenzene (DTBB) is used as a crystallized form from several organic solvents to study the effect of solvents and crystallization conditions on its habit. It is also used as a solvent and an intermediate in the manufacture of other organic compounds for the finished products of such as curing agents, engineering plastics and cross-linking agents.
Solubility
Insoluble in water. Soluble in ethanol, benzene, carbon tetrachloride.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under normal temperatures and pressures. Keep away from oxidizing agents.
Chemical Identifiers
1012-72-2 | |
190.33 | |
OOWNNCMFKFBNOF-UHFFFAOYSA-N | |
13895 | |
CC(C)(C)C1=CC=C(C=C1)C(C)(C)C |
C14H22 | |
MFCD00008836 | |
1,4-di-tert-butylbenzene, p-di-tert-butylbenzene, benzene, p-di-tert-butyl, benzene, 1,4-bis 1,1-dimethylethyl, unii-55ptx4kh71, bis 1,1-dimethylethyl benzene, benzene, bis 1,1-dimethylethyl, 1,4-di-t-butylbenzene, acmc-2097up, 1,4-bis tert-butyl benzeneShow More | |
1,4-ditert-butylbenzene |
Specifications
1012-72-2 | |
0.985 | |
C14H22 | |
1617000 | |
Insoluble in water. Soluble in ethanol,benzene,carbon tetrachloride. | |
CC(C)(C)C1=CC=C(C=C1)C(C)(C)C | |
190.33 | |
190.33 | |
1,4-Di-tert-butylbenzene |
75°C to 79°C | |
236°C | |
MFCD00008836 | |
1,4-di-tert-butylbenzene, p-di-tert-butylbenzene, benzene, p-di-tert-butyl, benzene, 1,4-bis 1,1-dimethylethyl, unii-55ptx4kh71, bis 1,1-dimethylethyl benzene, benzene, bis 1,1-dimethylethyl, 1,4-di-t-butylbenzene, acmc-2097up, 1,4-bis tert-butyl benzene | |
OOWNNCMFKFBNOF-UHFFFAOYSA-N | |
1,4-ditert-butylbenzene | |
13895 | |
98% |
Safety and Handling
missing translation for 'dotInformation' : Hazard Class: 9; Packaging Group: III
missing translation for 'einecsNumber' : 213-790-9
missing translation for 'rtecsNumber' : CY8444000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.