missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Diaminobutane, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
1,4-Diaminobutane is used as a precursor in many biological systems and synthon for amido-ureas. It is involved in the synthesis of nylon 46 by reacting with adipic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,4-Diaminobutane is used as a precursor in many biological systems and synthon for amido-ureas. It is involved in the synthesis of nylon 46 by reacting with adipic acid.
Solubility
Soluble in ether and alcohol.
Notes
Incompatible with acids, strong oxidizing agents, aldehydes, acid anhydrides and acid chlorides.
1,4-Diaminobutane is used as a precursor in many biological systems and synthon for amido-ureas. It is involved in the synthesis of nylon 46 by reacting with adipic acid.
Solubility
Soluble in ether and alcohol.
Notes
Incompatible with acids, strong oxidizing agents, aldehydes, acid anhydrides and acid chlorides.
Specifications
Specifications
Melting Point | 25°C to 28°C |
Density | 0.877 |
Boiling Point | 158°C to 160°C |
Flash Point | 63°C (145°F) |
Linear Formula | H2N(CH2)4NH2 |
Refractive Index | 1.4569 |
Quantity | 100 g |
UN Number | UN2928 |
Beilstein | 605282 |
Sensitivity | Air sensitive; Hygroscopic |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'