Learn More
1,4-Diaminobutane, 99%, Thermo Scientific Chemicals
£22.10 - £489.00
Chemical Identifiers
CAS | 110-60-1 |
---|---|
Molecular Formula | C4H12N2 |
Molecular Weight (g/mol) | 88.15 |
MDL Number | MFCD00008235 |
InChI Key | KIDHWZJUCRJVML-UHFFFAOYSA-N |
Synonym | 1,4-diaminobutane, putrescine, 1,4-butanediamine, tetramethylenediamine, butylenediamine, putrescin, 1,4-butylenediamine, tetramethyldiamine, 1,4-tetramethylenediamine, putrescina |
PubChem CID | 1045 |
ChEBI | CHEBI:17148 |
IUPAC Name | butane-1,4-diamine |
SMILES | C(CCN)CN |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10386410
|
Thermo Scientific Acros
112120250 |
25 g | Glass bottle |
£22.10
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10316130
|
Thermo Scientific Acros
112121000 |
100 g | Glass bottle |
£42.40
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10326130
|
Thermo Scientific
112125000 |
500 g | Glass bottle |
£126.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10267363
|
Thermo Scientific Acros
112120025 |
2.5 kg | Plastic bottle |
£489.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
110-60-1 | |
88.15 | |
KIDHWZJUCRJVML-UHFFFAOYSA-N | |
1045 | |
butane-1,4-diamine |
C4H12N2 | |
MFCD00008235 | |
1,4-diaminobutane, putrescine, 1,4-butanediamine, tetramethylenediamine, butylenediamine, putrescin, 1,4-butylenediamine, tetramethyldiamine, 1,4-tetramethylenediamine, putrescina | |
CHEBI:17148 | |
C(CCN)CN |
Specifications
110-60-1 | |
100.0 | |
Colorless to Yellow | |
158.0°C to 160.0°C | |
Authentic | |
C4H12N2 | |
1.4560 to 1.4610 | |
04,264 | |
0.877 | |
Solubility in water: 40g/L (20°C). | |
C(CCN)CN | |
88.15 | |
CHEBI:17148 | |
99% | |
1, 4-Diaminobutane, 99% |
98.5 | |
27.0°C | |
0.8770g/mL | |
45°C | |
98.5% min. (GC) | |
H2N(CH2)4NH2 | |
MFCD00008235 | |
15,8058 | |
1,4-diaminobutane, putrescine, 1,4-butanediamine, tetramethylenediamine, butylenediamine, putrescin, 1,4-butylenediamine, tetramethyldiamine, 1,4-tetramethylenediamine, putrescina | |
KIDHWZJUCRJVML-UHFFFAOYSA-N | |
butane-1,4-diamine | |
1045 | |
88.15 | |
Liquid or Low Melting Solid |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-782-3
missing translation for 'rtecsNumber' : EJ6800000
missing translation for 'tsca' : TSCA
- 1,4-Diaminobutane
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H330-Fatal if inhaled.
- H311-Toxic in contact with skin.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only