missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Dibromobutane, 98+%, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
10 g
100 g
500 g
Description
1,4-Dibromobutane is used as an intermediate involved in the synthesis of active pharmaceutical ingredient and other organic compounds. It is also used in the investigation of metabolism of two halopropanes such as 1,3-dichloropropane and 2,2-dichloropropane. Further, it acts as a reagent to prepare diazadioxa oxovanadium(IV) macrocyclic complexes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,4-Dibromobutane is used as an intermediate involved in the synthesis of active pharmaceutical ingredient and other organic compounds. It is also used in the investigation of metabolism of two halopropanes such as 1,3-dichloropropane and 2,2-dichloropropane. Further, it acts as a reagent to prepare diazadioxa oxovanadium(IV) macrocyclic complexes.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents and strong bases.
1,4-Dibromobutane is used as an intermediate involved in the synthesis of active pharmaceutical ingredient and other organic compounds. It is also used in the investigation of metabolism of two halopropanes such as 1,3-dichloropropane and 2,2-dichloropropane. Further, it acts as a reagent to prepare diazadioxa oxovanadium(IV) macrocyclic complexes.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications
Specifications
Melting Point | -20°C |
Density | 1.82 |
Boiling Point | 194°C to 196°C |
Flash Point | 131°C (267°F) |
Linear Formula | Br(CH2)4Br |
Refractive Index | 1.5186 |
Quantity | 500 g |
Beilstein | 1071199 |
Solubility Information | Immiscible with water. |
Formula Weight | 215.93 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'