Learn More
1,4-Dihydroxyanthraquinone, 95%, Thermo Scientific Chemicals
£36.50 - £343.00
Chemical Identifiers
CAS | 81-64-1 |
---|---|
Molecular Formula | C14H8O4 |
Molecular Weight (g/mol) | 240.214 |
MDL Number | MFCD00001209 |
InChI Key | GUEIZVNYDFNHJU-UHFFFAOYSA-N |
Synonym | quinizarin, 1,4-dihydroxyanthraquinone, quinizarine, chinizarin, solvent orange 86, 1,4-dihydroxyanthrachinon, smoke orange r, 1,4-dihydroxy-9,10-anthraquinone, macrolex orange gg, 9,10-anthracenedione, 1,4-dihydroxy |
PubChem CID | 6688 |
ChEBI | CHEBI:37487 |
IUPAC Name | 1,4-dihydroxyanthracene-9,10-dione |
SMILES | C1=CC=C2C(=C1)C(=O)C3=C(C=CC(=C3C2=O)O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11404483
|
Thermo Scientific Alfa Aesar
A11010.30 |
250 g |
£36.50
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11494473
|
Thermo Scientific Alfa Aesar
A11010.0B |
1000 g |
£95.50
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11414483
|
Thermo Scientific Alfa Aesar
A11010.0I |
5000 g | N/A | ||||||
Description
1,4-Dihydroxyanthraquinone is used to color gasoline and some heating oils. It is used as an intermediate for the synthesis of indanthrene- and alizarin-derived dyes. It is also used to form lake pigments with calcium, barium and lead. It may be used in the synthesis of cyclopentanoids by cyclization of α,β-unsaturated aldehyde with 1, 4 1,4-Dihydroxyanthraquinone. It has been studied as a long range emissive ratiometric fluorescent probe for live cell imaging.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,4-Dihydroxyanthraquinone is used to color gasoline and some heating oils. It is used as an intermediate for the synthesis of indanthrene- and alizarin-derived dyes. It is also used to form lake pigments with calcium, barium and lead. It may be used in the synthesis of cyclopentanoids by cyclization of α,β-unsaturated aldehyde with 1, 4 1,4-Dihydroxyanthraquinone. It has been studied as a long range emissive ratiometric fluorescent probe for live cell imaging.
Solubility
Soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
81-64-1 | |
240.214 | |
GUEIZVNYDFNHJU-UHFFFAOYSA-N | |
6688 | |
1,4-dihydroxyanthracene-9,10-dione |
C14H8O4 | |
MFCD00001209 | |
quinizarin, 1,4-dihydroxyanthraquinone, quinizarine, chinizarin, solvent orange 86, 1,4-dihydroxyanthrachinon, smoke orange r, 1,4-dihydroxy-9,10-anthraquinone, macrolex orange gg, 9,10-anthracenedione, 1,4-dihydroxy | |
CHEBI:37487 | |
C1=CC=C2C(=C1)C(=O)C3=C(C=CC(=C3C2=O)O)O |
Specifications
81-64-1 | |
∼450°C | |
C14H8O4 | |
1914036 | |
quinizarin, 1,4-dihydroxyanthraquinone, quinizarine, chinizarin, solvent orange 86, 1,4-dihydroxyanthrachinon, smoke orange r, 1,4-dihydroxy-9,10-anthraquinone, macrolex orange gg, 9,10-anthracenedione, 1,4-dihydroxy | |
GUEIZVNYDFNHJU-UHFFFAOYSA-N | |
1,4-dihydroxyanthracene-9,10-dione | |
6688 | |
240.21 | |
1,4-Dihydroxyanthraquinone |
195°C to 198°C | |
222°C (431°F) | |
MFCD00001209 | |
14,8064 | |
Soluble in water. | |
C1=CC=C2C(=C1)C(=O)C3=C(C=CC(=C3C2=O)O)O | |
240.214 | |
CHEBI:37487 | |
95% |
Safety and Handling
missing translation for 'dotInformation' : Hazard Class: 9; Packaging Group: III
missing translation for 'einecsNumber' : 201-368-7
missing translation for 'rtecsNumber' : CB6600000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only