missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1,4-Dimethyl-2-piperazinyl)methanol, 95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Specifications
Specifications
CAS Min % | 95.0 |
Assay Percent Range | 95% |
Packaging | Amber glass bottle |
Quantity | 1 g |
Formula Weight | 144.22 |
Percent Purity | 95% |
Chemical Name or Material | (1, 4-Dimethyl-2-piperazinyl)methanol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction