Learn More
Chemical Identifiers
CAS | 502-97-6 |
---|---|
Molecular Formula | C4H4O4 |
Molecular Weight (g/mol) | 116.07 |
MDL Number | MFCD00081108 |
InChI Key | RKDVKSZUMVYZHH-UHFFFAOYSA-N |
Synonym | glycolide, glycollide, unii-yrz676pgu6, p-dioxane-2,5-dione, yrz676pgu6, acmc-1aybf, 1,4-dioxan-2,5-dione, 2,5-dioxo-1,4-dioxane, p-dioxane-2,5-dione 8ci |
PubChem CID | 65432 |
IUPAC Name | 1,4-dioxane-2,5-dione |
SMILES | O=C1COC(=O)CO1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10223612
|
Thermo Scientific Maybridge
SB01616EA |
10 g | Amber Glass Bottle |
£46.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10101132
|
Thermo Scientific Maybridge
SB01616EE |
50 g | Amber Glass Bottle |
£186.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
502-97-6 | |
116.07 | |
RKDVKSZUMVYZHH-UHFFFAOYSA-N | |
65432 | |
O=C1COC(=O)CO1 |
C4H4O4 | |
MFCD00081108 | |
glycolide, glycollide, unii-yrz676pgu6, p-dioxane-2,5-dione, yrz676pgu6, acmc-1aybf, 1,4-dioxan-2,5-dione, 2,5-dioxo-1,4-dioxane, p-dioxane-2,5-dione 8ci | |
1,4-dioxane-2,5-dione |
Specifications
502-97-6 | |
97% | |
MFCD00081108 | |
RKDVKSZUMVYZHH-UHFFFAOYSA-N | |
1,4-dioxane-2,5-dione | |
65432 | |
97% |
97.0 | |
C4H4O4 | |
glycolide, glycollide, unii-yrz676pgu6, p-dioxane-2,5-dione, yrz676pgu6, acmc-1aybf, 1,4-dioxan-2,5-dione, 2,5-dioxo-1,4-dioxane, p-dioxane-2,5-dione 8ci | |
O=C1COC(=O)CO1 | |
116.07 | |
116.07 | |
1, 4-Dioxane-2, 5-dione |
Safety and Handling
GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.